missing translation for 'onlineSavingsMsg'
Learn More

CD107b (LAMP-2), Alexa Fluor 488, clone: eBioABL-93 (ABL-93), eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 53-1072-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15372000

  • £62.63 / 25µg


Estimated Shipment date
from Supplier 26-03-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The eBioABL-93 monoclonal antibody reacts with mouse CD107b, also known as lysosomal-associated membrane protein-2 (LAMP-2). CD107b is a highly glycosylated protein of approximately 120kDa that is expressed intracellularly in lysosomal membranes. It is also transiently expressed on the surface of cytolytic T cells during degranulation, but to a lesser extent than CD107a. It has been reported that CD107b may be identical to the mouse Mac-3 antigen.Applications Reported: This eBioABL-93 (ABL-93) antibody has been reported for use in intracellular staining followed by flow cytometric analysis. It is also reported for use in surface staining in a flow cytometric based degranulation assay.Applications Tested: This eBioABL-93 (ABL-93) antibody has been tested by intracellular staining and flow cytometric analysis of thioglycollate elicited peritoneal macrophages. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD107b (LAMP-2)
eBioABL-93 (ABL-93)
Alexa Fluor 488
Conjugated
Rat
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry (Intracellular Staining)
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
LAMP2, lysosomal-associated membrane protein 2
IgG2a
RUO
Primary
Murine
Videos