missing translation for 'onlineSavingsMsg'
Learn More

CD107b (LAMP-2), PE, clone: eBioH4B4 (H4B4), eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-1078-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15536856

  • £147.88 / 100 tests


Estimated Shipment date
from Supplier 26-06-2018
Add to basket

Description

Description

Description: The eBioH4B4 monoclonal antibody reacts with human CD107b, also known as lysosomal-associated membrane protein-2 (LAMP-2). CD107b is a highly glycosylated, type I transmembrane protein of approximately 105 kDa. It is expressed intracellularly in lysosomal/endosomal membranes in nearly all cells. It is also expressed on the surface of degranulating T cells (to a lesser extent than CD107a) and activated platelets as well as some cancer cells. In humans, mutations in CD107b results in a lysosomal glycogen storage disorder, known as Danon disease.Applications Reported: This eBioH4B4 (H4B4) antibody has been reported for use in intracellular staining followed by flow cytometric analysis.Applications Tested: This eBioH4B4 (H4B4) antibody has been pre-titrated and tested by intracellular staining and flow cytometric analysis of Jurkat cells using the Intracellular Fixation & Permeabilization Buffer Set (cat. 88-8824) and protocol. Please refer to Best Protocols: Protocol A: Two step protocol for (cytoplasmic) intracellular proteins located under the Resource Tab online. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD107b (LAMP-2)
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P13473
RP23-193O17.2, CD107b, Lamp-2, Lamp-2a, Lamp-2b, Lamp-2c, Mac3
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
eBioH4B4 (H4B4)
PE
LAMP2
Conjugated
LAMP2
IgG1, kappa
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
3920
Videos

For Research Use Only.