missing translation for 'onlineSavingsMsg'
Learn More

CD107b (LAMP-2), PE, clone: eBioH4B4 (H4B4), eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-1078-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15526856

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

The eBioH4B4 monoclonal antibody reacts with human CD107b, also known as lysosomal-associated membrane protein-2 (LAMP-2). CD107b is a highly glycosylated, type I transmembrane protein of approximately 105 kDa. It is expressed intracellularly in lysosomal/endosomal membranes in nearly all cells. It is also expressed on the surface of degranulating T cells (to a lesser extent than CD107a) and activated platelets as well as some cancer cells. In humans, mutations in CD107b results in a lysosomal glycogen storage disorder, known as Danon disease.
Specifications

Specifications

CD107b (LAMP-2)
eBioH4B4 (H4B4)
PE
Conjugated
Murine
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
5μL (0.5μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
lysosomal-associated membrane protein 2
IgG1, kappa
RUO
Primary
Human
Videos

For Research Use Only.