missing translation for 'onlineSavingsMsg'
Learn More

CD107b (LAMP-2) Mouse anti-Human, eFluor® 660, Clone: eBioH4B4 (H4B4), eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 50-1078-41

Code : NEW

Additional Details : Weight : 0.23750kg

Product Code. 15579896

  • £67.75 / 25 tests

Estimated Shipment
04-11-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Lysosomes are membrane-bound vacuoles which play a critical role in cellular metabolism. These organelles participate in endocytosis, the synthesis and assembly of acid hydrolases, act as sites for digestion of foreign materials and for specialized autolytic cellular processes. Membrane-associated glycoproteins have an important role in normal lysosomal function. Prior to posttranslational modification, Lysosome Associated Membrane Protein 2 (LAMP 2) is an ∽45 kDa polypeptide. Mature, functional LAMP 2 is extensively glycosylated with a variety of different N-linked and O-linked oligosaccharides with a total molecular weight of ∽100-110 kDa. LAMP 2 has a structure comprised of a large amino-terminal intra-lysosomal domain, a hydrophobic trans-membrane domain, and a short carboxyl-terminal cytoplasmic tail.
Specifications

Specifications

CD107b (LAMP-2)
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P13473
RP23-193O17.2, CD107b, Lamp-2, Lamp-2a, Lamp-2b, Lamp-2c, Mac3
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
eBioH4B4 (H4B4)
eFluor™ 660
LAMP2
Liquid
LAMP2
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
3920
Videos
Documents

For Research Use Only.