missing translation for 'onlineSavingsMsg'
Learn More

CD107a (LAMP-1), Biotin, clone: eBio1D4B (1D4B), eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-1071-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15279679

  • £56.91 / 50µg


Estimated Shipment date
from Supplier 04-04-2018
Add to basket

Description

Description

Description: The eBio1D4B monoclonal antibody reacts with mouse CD107a, also known as lysosomal-associated membrane protein-1 (LAMP-1). CD107a is a type I, lysosomal membrane protein that is extensively glycosylated. It is expressed constitutively in the late endosomes-lysosomes in all cells. CD107a is also transiently expressed on the cell surface of degranulating cytolytic T cells. Additionally, CD107a has been implicated in a variety of cellular functions including cancer metastasis and is also a marker for lysosomal storage disorders.Applications Reported: This eBio1D4B (1D4B) antibody has been reported for use in intracellular staining followed by flow cytometric analysis. It has also been reported for use in surface staining in a flow cytometric based degranulation assay.Applications Tested: This eBio1D4B (1D4B) antibody has been tested by intracellular staining and flow cytometric analysis of thioglycolate-elicited peritoneal exudate cells. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD107a (LAMP-1)
eBio1D4B (1D4B)
Biotin
Conjugated
Rat
50μg
Store at 2-8°C. Do not freeze.
Monoclonal
Flow Cytometry (Intracellular Staining)
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
lysosomal-associated membrane protein 1
IgG2a, kappa
RUO
Primary
Murine
Videos

For Research Use Only.