Learn More
CD107a (LAMP-1) Monoclonal Antibody (eBioH4A3), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£100.00 - £279.00
Specifications
Antigen | CD107a (LAMP-1) |
---|---|
Clone | eBioH4A3 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | PerCP-eFluor 710 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15519176
|
Affymetrix eBioscience
46-1079-42 |
100 Tests |
£279.00
100 tests |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15509176
|
Affymetrix eBioscience
46-1079-41 |
25 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The eBioH4A3 monoclonal antibody reacts with human CD107a, also known as lysosomal-associated membrane protein-1 (LAMP-1). CD107a is a highly glycosylated protein of approximately 110kDa. It is predomitly expressed intracellularly in the lysosomal/endosomal membrane in nearly all cells. CD107a is transiently expressed on the cell surface of degranulating cytolytic T cells, and is also upregulated on the surface of activated platelets and some cancer cells. Applications Reported: This eBioH4A3 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This eBioH4A3 antibody has been pre-titrated and tested by intracellular staining and flow cytometric analysis of Jurkat cells using the Intracellular Fixation & Permeabilization Buffer Set (cat. 88-8824) and protocol. Please refer to Best Protocols: Protocol A: Two step protocol for (cytoplasmic) intracellular proteins located under the Resources Tab online. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. PerCP-eFluor™ 710 emits at 710 nm and is excited with the blue laser (488 nm); it can be used in place of PerCP-Cyanine5.5.
CD107a (LAMP-1) Monoclonal antibody specifically detects CD107a (LAMP-1) in Human samples. It is validated for Flow Cytometry.Specifications
CD107a (LAMP-1) | |
Flow Cytometry | |
PerCP-eFluor 710 | |
Mouse | |
Human | |
3916 | |
IgG1 κ | |
Affinity chromatography | |
LAMP1 |
eBioH4A3 | |
Monoclonal | |
Liquid | |
RUO | |
P11279 | |
LAMP1 | |
Primary | |
Antibody |
For Research Use Only.