Learn More
CD107a (LAMP-1), PE-eFluor 610, clone: eBioH4A3, eBioscience™
Mouse Monoclonal Antibody
Brand:  Affymetrix eBioscience 61-1079-41
Code : NEW
Additional Details : Weight : 0.23750kg
 View more versions of this product
Product Code. 15520457
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Disclaimers:
For Research Use Only.
Description
Description
The eBioH4A3 monoclonal antibody reacts with human CD107a, also known as lysosomal-associated membrane protein-1 (LAMP-1). CD107a is a highly glycosylated protein of approximately 110kDa. It is predominantly expressed intracellularly in the lysosomal/endosomal membrane in nearly all cells. CD107a is transiently expressed on the cell surface of degranulating cytolytic T cells, and is also upregulated on the surface of activated platelets and some cancer cells.
Specifications
Specifications
CD107a (LAMP-1)
eBioH4A3
PE-eFluor 610
Conjugated
Murine
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
Flow Cytometry (Intracellular Staining)
5μL (0.25μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
LAMP1, lysosomal-associated membrane protein 1
IgG1