missing translation for 'onlineSavingsMsg'
Learn More

CD106 (VCAM-1), clone: 429, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-1061-85

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15287157

  • £137.64 / 500µg


EU Stock 1
Add to basket

Description

Description

Description: The 429 monoclonal antibody reacts with mouse CD106 (Vascular Cell Adhesion Molecule-1, VCAM-1), a 110kDa transmembrane glycoprotein expressed by myeloid lineage and bone marrow stromal cells. Endothelial cells constitutively express low levels of CD106 and upregulate it upon cytokine stimulation. CD106 binds to integrin alpha4beta1 (VLA-4, CD49d/CD29) and Integrin a4b7 (LPAM-1) and these interactions in the bone marrow and thymus are important for early lymphocyte and myeloid development. Cytokine-mediated upregulation of CD106 on endothelial cells suggests a role for this antigen in the inflammatory response.Applications Reported: This 429 antibody has been reported for use in flow cytometric analysis, immunoprecipitation, and immunohistochemical staining of frozen tissue sections (IHC-F). It has also been reported in blocking of CD106 in functional studies. (Please use Functional Grade purified 429, cat. 16-1061, in functional assays.).Applications Tested: The 429 antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD106 (VCAM-1)
Monoclonal
0.5mg/mL
PBS with 0.09% sodium azide; pH 7.2
P29533
CD106, DKFZp779G2333, INCAM-100, MGC99561, CD106 antigen, vascular cell adhesion protein 1
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Immunoprecipitation
429
Unconjugated
Vcam1
Purified
Vcam1
IgG2a, kappa
500μg
Store at 2-8°C.
Primary
22329
Videos

For Research Use Only.