missing translation for 'onlineSavingsMsg'
Learn More

CD106 (VCAM-1), FITC, clone: STA, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-1069-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15516476

  • £62.50 / 25 tests


Estimated Shipment date
from Supplier 19-02-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The STA monoclonal antibody reacts with human CD106 (Vascular Cell Adhesion Molecule-1, VCAM-1), a 110kDa transmembrane glycoprotein expressed by myeloid lineage and bone marrow stromal cells. Endothelial cells constitutively express low levels of CD106 and upregulate it upon cytokine stimulation. CD106 binds to Integrin alpha4beta1 (VLA-4, CD49d/CD29) and Integrin alpha4beta7 (LPAM-1). Cytokine-mediated upregulation of CD106 on endothelial cells suggests a role for this antigen in the inflammatory response.Applications Reported: This STA antibody has been reported for use in flow cytometric analysis.Applications Tested: This STA antibody has been pre-titrated and tested by flow cytometric analysis of stimulated HUVEC. This can be used at 5 μL (1 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD106 (VCAM-1)
STA
FITC
Conjugated
Murine
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
5μL (1μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
vascular cell adhesion molecule-1
IgG1
RUO
Primary
Human
Videos