missing translation for 'onlineSavingsMsg'
Learn More

CD105 (Endoglin), eFluor 450, clone: MJ7/18, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 48-1051-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15381740

  • £62.06 / 25µg


EU Stock 3
Add to basket

Description

Description

The MJ7/18 monoclonal antibody reacts with the mouse CD105 molecule, also known as Endoglin. This 90kDa disulfide-linked homodimer is expressed by vascular endothelial cells. It is suggested that CD105 functions in adhesion and embryonic angiogenesis.
Specifications

Specifications

CD105
MJ7/18
eFluor 450
Conjugated
IgG2a, kappa
RUO
Primary
Murine
Flow Cytometry
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Rat
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Videos

For Research Use Only.