Learn More
CD105 (Endoglin) Monoclonal Antibody (MJ7/18), APC, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 17-1051-80
Code : NEW
Additional Details : Weight : 0.09500kg
Description
Description: The MJ7/18 monoclonal antibody reacts with the mouse CD105 molecule, also known as Endoglin. This 90 kDa disulfide-linked homodimer is expressed by vascular endothelial cells. It is suggested that CD105 functions in adhesion and embryonic angiogenesis. Applications Reported: This MJ7/18 antibody has been reported for use in flow cytometric analysis. Applications Tested: This MJ7/18 antibody has been tested by flow cytometric analysis of bEnd.3 cells. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
CD105 (Endoglin) Monoclonal antibody specifically detects CD105 (Endoglin) in Mouse samples. It is validated for Flow Cytometry.Specifications
CD105 (Endoglin) | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Q63961 | |
ENG | |
Affinity chromatography | |
RUO | |
13805 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
MJ7/18 | |
APC | |
ENG | |
AI528660; AI662476; CD105; CD105 antigen; cell surface MJ7/18 antigen; Edg; END; Endo; Endoglin; endoglin (Osler-Rendu-Weber syndrome 1); Eng; HHT1; ORW; ORW1; S-endoglin; transmembrane glycoprotein | |
Rat | |
25 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a κ |
For Research Use Only.