Learn More
CD105 (Endoglin) Monoclonal Antibody (SN6), PE-Cyanine7, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£97.00 - £246.00
Specifications
Antigen | CD105 (Endoglin) |
---|---|
Clone | SN6 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15528456
|
Affymetrix eBioscience
25-1057-41 |
25 Tests |
£97.00
25 tests |
Estimated Shipment: 09-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15538456
|
Affymetrix eBioscience
25-1057-42 |
100 Tests |
£246.00
100 tests |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The SN6 monoclonal antibody reacts with human CD105, also known as Endoglin. CD105, an approximately 90 kDa disulfide-linked homodimer is expressed by vascular endothelial cells and some bone marrow cells and activated macrophages. It is suggested that CD105 functions in adhesion and embryonic angiogenesis. Applications Reported: This SN6 antibody has been reported for use in flow cytometric analysis. Applications Tested: This SN6 antibody has been pre-titrated and tested by flow cytometric analysis of the U937 cell line. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Light sensitivity: This tandem dye is sensitive photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488-561 nm; Emission: 775 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.
CD105 (Endoglin) Monoclonal antibody specifically detects CD105 (Endoglin) in Human samples. It is validated for Flow Cytometry.Specifications
CD105 (Endoglin) | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
AI528660; AI662476; CD105; CD105 antigen; cell surface MJ7/18 antigen; Edg; END; Endo; Endoglin; endoglin (Osler-Rendu-Weber syndrome 1); Eng; HHT1; ORW; ORW1; S-endoglin; transmembrane glycoprotein | |
ENG | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
ENG |
SN6 | |
Flow Cytometry | |
PE-Cyanine7 | |
Mouse | |
Human | |
P17813 | |
2022 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.