missing translation for 'onlineSavingsMsg'
Learn More

CD105 (Endoglin) Mouse anti-Human, PE, Clone: SN6, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-1057-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15566846

  • £68.00 / 25 tests

Estimated Shipment date
from Supplier 06-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CD105 or Endoglin is a Type I transmembrane protein, which is highly expressed on human vascular endothelial cells. It exists on an O- and N-glycosylated homodimer. Up regulation of endoglin expression has been demonstrated in tumor vasculature and proliferating cells, suggesting that it is a proliferation associated endothelial marker. CD105 binds to TGF beta 1 and 3 with high affinity but not to TGF beta 2.
Specifications

Specifications

CD105 (Endoglin)
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P17813
Endoglin
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
SN6
PE
ENG
Liquid
ENG
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
2022
Videos
Documents

For Research Use Only.