Learn More
CD103 Monoclonal Antibody (LF61), TRI-COLOR™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MHCD10306
Code : 50
Additional Details : Weight : 0.50000kg
Description
The TRI-COLOR® conjugate is a tandem R-phycoerythrin (PE)-Cy5® conjugate that permits simultaneous multicolor labeling and detection of multiple targets with excitation by a single excitation source-the 488 nm line of the argon-ion laser.
CD103 Monoclonal antibody specifically detects CD103 in Human samples. It is validated for Flow Cytometry.Specifications
CD103 | |
Monoclonal | |
TRI-COLOR | |
Itgae | |
A530055J10; alpha-E1; alpha-M290; aM290; antigen CD103; antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide; CD103; DC; DCs; HML-1 antigen; human mucosal lymphocyte antigen 1, alpha polypeptide; HUMINAE; integrin alpha E; integrin alpha E, epithelial-associated; integrin alpha E1, epithelial-associated; integrin alpha M290; Integrin alpha-E; Integrin alpha-E heavy chain; Integrin alpha-E light chain; Integrin alpha-IEL; integrin subunit alpha E; integrin, alpha E; integrin, alpha E (antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide); integrin, alpha E, epithelial-associated; Intregrin alpha E2; Itgae; MGC141996; Mucosal lymphocyte 1 antigen | |
Mouse | |
purified | |
ASR | |
3682 | |
4° C, store in dark | |
Liquid |
Flow Cytometry | |
LF61 | |
PBS with sucrose and 0.1% sodium azide | |
P38570 | |
Itgae | |
Human CD103 | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |