missing translation for 'onlineSavingsMsg'
Learn More

CD103 (Integrin alpha E), PE, clone: OX62, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-1030-82

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15268629

  • £117.97 / 100µg


EU Stock 2
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The OX62 monoclonal antibody reacts with rat CD103, also known as alphaE2 integrin. CD103 was originally identified on rat veiled dendritic cells, and is expressed by dendritic epidermal T cells in the skin, intraepithelial lymphocytes in the small intestine and by dendritic cells associated with gammadelta T cells. Integrins are a family of heterodimeric proteins which contain non-covalently associated alpha and beta transmembrane glycoproteins. Integrins function as receptors for extracellular matrix and cell-cell adhesion molecules.Applications Reported: This OX62 antibody has been reported for use in flow cytometric analysis.Applications Tested: This OX62 antibody has been tested by flow cytometric analysis of rat splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD103 (Integrin alpha E)
OX62
PE
Conjugated
Murine
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Integrin aE, ITGAE
IgG1
RUO
Primary
Rat
Videos