missing translation for 'onlineSavingsMsg'
Learn More

CD103 (Integrin alpha E), PE, clone: OX62, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-1030-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15258629

  • £55.74 / 25µg


Estimated Shipment date
from Supplier 03-07-2018
Add to basket

Description

Description

Description: The OX62 monoclonal antibody reacts with rat CD103, also known as alphaE2 integrin. CD103 was originally identified on rat veiled dendritic cells, and is expressed by dendritic epidermal T cells in the skin, intraepithelial lymphocytes in the small intestine and by dendritic cells associated with gammadelta T cells. Integrins are a family of heterodimeric proteins which contain non-covalently associated alpha and beta transmembrane glycoproteins. Integrins function as receptors for extracellular matrix and cell-cell adhesion molecules.Applications Reported: This OX62 antibody has been reported for use in flow cytometric analysis.Applications Tested: This OX62 antibody has been tested by flow cytometric analysis of rat splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD103
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Conjugated
Itgae
IgG1
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
83577
Flow Cytometry
OX62
PE
Itgae
ITGAE, HUMINAE
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Rat
Videos

For Research Use Only.