missing translation for 'onlineSavingsMsg'
Learn More

CD103 (Integrin alpha E), PE, clone: B-Ly7, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-1038-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15556846

  • £150.07 / 100 tests


Estimated Shipment date
from Supplier 19-01-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The B-Ly7 monoclonal antibody reacts with human CD103, the alpha E integrin. CD103 non-covalently associates with integrin beta 7. CD103 is expressed mainly on intraepithelial lymphocytes and a small subset of peripheral lymphocytes. CD103 is also expressed by hairy cell leukemia (HCL) and by some chronic B cell lymphocytic leukemias. In vitro stimulation of human T cells with mitogens induces upregulation of CD103. Epithelial cell antigen, E-cadherin, binds to CD103 and mediates homing of lymphocytes to the intestinal epithelium.Applications Reported: B-Ly7 has been reported for use in flow cytometric analysis.Applications Tested: This B-Ly7 antibody has been pre-titrated and tested by flow cytometric analysis of stimulated normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD103 (Integrin alpha E)
B-Ly7
PE
Conjugated
Murine
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
5μL (0.25 μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
ITGAE
IgG1
RUO
Primary
Human
Videos