missing translation for 'onlineSavingsMsg'
Learn More

CD103 (Integrin alpha E), clone: B-Ly7, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-1038-80

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15247147

  • £37.33 / 25µg


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

Description: The B-Ly7 monoclonal antibody reacts with human CD103, the alpha E integrin. CD103 non-covalently associates with integrin beta 7. CD103 is expressed mainly on intraepithelial lymphocytes and a small subset of peripheral lymphocytes. CD103 is also expressed by hairy cell leukemia (HCL) and by some chronic B cell lymphocytic leukemias. In vitro stimulation of human T cells with mitogens induces upregulation of CD103. Epithelial cell antigen, E-cadherin, binds to CD103 and mediates homing of lymphocytes to the intestinal epithelium.Applications Reported: B-Ly7 has been reported for use in flow cytometric analysis, and immunohistochemical staining of frozen tissue sections.Applications Tested: The B-Ly7 antibody has been tested by flow cytometric analysis of stimulated normal human peripheral blood cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD103 (Integrin alpha E)
Monoclonal
0.5mg/mL
PBS with 0.09% sodium azide; pH 7.2
P38570
ITGAE, HUMINAE
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Frozen)
B-Ly7
Unconjugated
ITGAE
Purified
ITGAE
IgG1, kappa
25μg
Store at 2-8°C.
Primary
3682
Videos

For Research Use Only.