missing translation for 'onlineSavingsMsg'
Learn More

CD103 (Integrin alpha E), APC, clone: B-Ly7, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-1038-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15507986

  • £156.06 / 100 tests


EU Stock 4
Add to basket

Description

Description

Description: The B-Ly7 monoclonal antibody reacts with human CD103, the alpha E integrin. CD103 non-covalently associates with integrin beta 7. CD103 is expressed mainly on intraepithelial lymphocytes and a small subset of peripheral lymphocytes. CD103 is also expressed by hairy cell leukemia (HCL) and by some chronic B cell lymphocytic leukemias. In vitro stimulation of human T cells with mitogens induces upregulation of CD103. Epithelial cell antigen, E-cadherin, binds to CD103 and mediates homing of lymphocytes to the intestinal epithelium.Applications Reported: This B-Ly7 antibody has been reported for use in flow cytometric analysis.Applications Tested: This B-Ly7 antibody has been pre-titrated and tested by flow cytometric analysis of stimulated normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD103 (Integrin alpha E)
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P38570
ITGAE, HUMINAE
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
B-Ly7
APC
ITGAE
Conjugated
ITGAE
IgG1, kappa
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
3682
Videos

For Research Use Only.