missing translation for 'onlineSavingsMsg'
Learn More

CD103 (Integrin alpha E) Mouse anti-Human, APC, Clone: B-Ly7, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-1038-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15597976

  • £71.75 / 25 tests

Estimated Shipment
29-10-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The CD103 antigen is a 160-130-105 kDa molecule which is expressed on mucosa-associated T lymphocytes and activated cells and on subet of TGF beta -1 cells. This human intra epithelial lymphocyte marker is the E integrin chain associated with the b7 subunit. CD103 antibody stains a few bone marrow cells and, rarely, peripheral blood lymphocytes. CD103 is expressed by 0.5 to 2% of resting lymphocytes in peripheral blood and lymphoid organs.
Specifications

Specifications

CD103 (Integrin alpha E)
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P38570
ITGAE, HUMINAE
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
B-Ly7
APC
ITGAE
Liquid
ITGAE
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
3682
Videos
Documents

For Research Use Only.