missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD103 (Integrin alpha E) Monoclonal Antibody (2E7), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Armenian Hamster Monoclonal Antibody
£118.00 - £278.00
Specifications
Antigen | CD103 (Integrin alpha E) |
---|---|
Clone | 2E7 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15569166
|
Affymetrix eBioscience
46-1031-80 |
25 μg |
£118.00
25µg |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15579166
|
Affymetrix eBioscience
46-1031-82 |
100 μg |
£278.00
100µg |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The CD103 antigen is a 160-130-105kDa molecule which is expressed on mucosa-associated T lymphocytes and activated cells and on subet of TGF beta -1 cells. This human intra epithelial lymphocyte marker is the E integrin chain associated with the b7 subunit. CD103 antibody stains a few bone marrow cells and, rarely, peripheral blood lymphocytes. CD103 is expressed by 0.5 to 2% of resting lymphocytes in peripheral blood and lymphoid organs.
CD103 (Integrin alpha E) Monoclonal antibody specifically detects CD103 (Integrin alpha E) in Mouse samples. It is validated for Flow Cytometry.Specifications
CD103 (Integrin alpha E) | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
A530055J10; alpha-E1; alpha-M290; aM290; antigen CD103; antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide; CD103; DC; DCs; HML-1 antigen; human mucosal lymphocyte antigen 1, alpha polypeptide; HUMINAE; integrin alpha E; integrin alpha E, epithelial-associated; integrin alpha E1, epithelial-associated; integrin alpha M290; Integrin alpha-E; Integrin alpha-E heavy chain; Integrin alpha-E light chain; Integrin alpha-IEL; integrin subunit alpha E; integrin, alpha E; integrin, alpha E (antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide); integrin, alpha E, epithelial-associated; Intregrin alpha E2; Itgae; MGC141996; Mucosal lymphocyte 1 antigen | |
Itgae | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Itgae |
2E7 | |
Flow Cytometry | |
PerCP-eFluor 710 | |
Armenian Hamster | |
Mouse | |
Q60677 | |
16407 | |
IgG | |
Affinity chromatography | |
Antibody |
For Research Use Only.