missing translation for 'onlineSavingsMsg'
Learn More

CD103 (Integrin alpha E), FITC, clone: 2E7, eBioscience™

Armenian Hamster Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-1031-82

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15288159

  • £105.77 / 100µg


EU Stock 7
Add to basket

Description

Description

Description: The 2E7 monoclonal antibody reacts with mouse CD103, the alphaE integrin. CD103 non-covalently associates with integrin beta7 and is expressed on all intraepithelial lymphocytes, a small subset of peripheral lymphocytes, and dendritic epidermal T cells (DEC). Epithelial cell antigen E-cadherin binds to CD103 and mediates homing of lymphocytes to the intestinal epithelium.Applications Reported: The 2E7 antibody has been reported for use in flow cytometric analysis.Applications Tested: The 2E7 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD103
Monoclonal
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Q60677
ITGAE, HUMINAE
Armenian Hamster
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
2.00E+07
FITC
Itgae
Conjugated
Itgae
IgG
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
16407
Videos

For Research Use Only.