Learn More
CD103 (Integrin alpha E) Monoclonal Antibody (2E7), eFluor™ 450, eBioscience™, Invitrogen™
Armenian Hamster Monoclonal Antibody
Brand: Affymetrix eBioscience 48-1031-80
Code : NEW
Additional Details : Weight : 0.09500kg
Description
Description: The 2E7 monoclonal antibody reacts with mouse CD103, the alphaE integrin. CD103 non-covalently associates with integrin beta7 and is expressed on all intraepithelial lymphocytes, a small subset of peripheral lymphocytes, and dendritic epidermal T cells (DEC). Epithelial cell antigen E-cadherin binds to CD103 and mediates homing of lymphocytes to the intestinal epithelium. Applications Reported: This 2E7 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 2E7 antibody has been tested by flow cytometric analysis of mouse lymph node cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. eFluor™ 450 is an alternative to Pacific Blue™. eFluor™ 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser. Filtration: 0.2 μm post-manufacturing filtered.
CD103 (Integrin alpha E) Monoclonal antibody specifically detects CD103 (Integrin alpha E) in Mouse samples. It is validated for Flow Cytometry.Specifications
CD103 (Integrin alpha E) | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Q60677 | |
Itgae | |
Affinity chromatography | |
RUO | |
16407 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
2E7 | |
eFluor 450 | |
Itgae | |
A530055J10; alpha-E1; alpha-M290; aM290; antigen CD103; antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide; CD103; DC; DCs; HML-1 antigen; human mucosal lymphocyte antigen 1, alpha polypeptide; HUMINAE; integrin alpha E; integrin alpha E, epithelial-associated; integrin alpha E1, epithelial-associated; integrin alpha M290; Integrin alpha-E; Integrin alpha-E heavy chain; Integrin alpha-E light chain; Integrin alpha-IEL; integrin subunit alpha E; integrin, alpha E; integrin, alpha E (antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide); integrin, alpha E, epithelial-associated; Intregrin alpha E2; Itgae; MGC141996; Mucosal lymphocyte 1 antigen | |
Armenian Hamster | |
25 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |
For Research Use Only.