missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICAM-2 Monoclonal Antibody (3C4), FITC, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen A15450
Code : NEW
Additional Details : Weight : 0.00280kg
Description
ICAM-2 Monoclonal antibody specifically detects ICAM-2 in Mouse samples. It is validated for Flow Cytometry.Specifications
ICAM-2 | |
Monoclonal | |
0.1 mg/mL | |
PBS with <0.1% sodium azide | |
P35330 | |
ICAM2 | |
Mouse CD102 from COS cells transfected with mouse ICAM-2 cDNA and BW5147 AKR/J mouse thymoma cell line | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a κ |
Flow Cytometry | |
3C4 | |
FITC | |
ICAM2 | |
CD102; Icam2; ICAM-2; Intercellular adhesion molecule 2; Ly-60; Lymphocyte function-associated AG-1 counter-receptor; RP23-418O11.10-001 | |
Rat | |
purified | |
RUO | |
15896 | |
4° C, store in dark | |
Liquid |