missing translation for 'onlineSavingsMsg'
Learn More

CD102 (ICAM-2) Rat anti-Mouse, Functional Grade, Clone: 3C4 (mIC2/4), eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 16-1021-85

Code : NEW

Additional Details : Weight : 0.09500kg

Product Code. 15587776

  • £185.00 / 500µg

Estimated Shipment date
from Supplier 02-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

001 ng/μg antibody, as determined by LAL assay. Aggregation: Less than 10%, as determined by HPLC. ICAM-2 is a member of the intercellular adhesion molecule (ICAM) family. All ICAM proteins are type I transmembrane glycoproteins, contain 2-9 immunoglobulin-like C2-type domains, and bind to the leukocyte adhesion LFA-1 protein. ICAM-2 may play a role in lymphocyte recirculation by blocking LFA-1-dependent cell adhesion. It mediates adhesive interactions important for antigen-specific immune response, NK-cell mediated clearance, lymphocyte recirculation, and other cellular interactions important for immune response and surveillance. Several transcript variants encoding the same protein have been found for this gene. Diseases associated with ICAM2 include Colon Carcinoma In Situ.
Specifications

Specifications

CD102 (ICAM-2)
Monoclonal
1 mg/mL
PBS with no preservative; pH 7.2
P35330
RP23-418O11.10-001, CD102, Icam-2, Ly-60, lymphocyte function-associated AG-1 counter-receptor, Intercellular adhesion molecule 2
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry, Functional Assay
3C4 (mIC2/4)
Functional Grade
Icam2
Liquid
Icam2
IgG2a, kappa
500 μg
4° C
Primary
15896
Videos
Documents

For Research Use Only.