Learn More
CD102 (ICAM-2) Monoclonal Antibody (3C4 (mIC2/4)), Alexa Fluor™ 488, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£186.00
Specifications
Antigen | CD102 (ICAM-2) |
---|---|
Clone | 3C4 (mIC2/4) |
Concentration | 0.5 mg/mL |
Applications | Flow Cytometry, Immunohistochemistry (Frozen) |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15342000
|
Affymetrix eBioscience
53-1021-82 |
100 μg |
£186.00
100µg |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The monoclonal antibody 3C4 (mIC2/4) recognizes mouse CD102, also known as intercellular adhesion molecule-2 (ICAM-2). CD102, a member of the Immunoglobulin superfamily of receptors with 2 extracellular Ig domains, binds LFA (CD11a/CD18) and Mac-1 (CD11b/CD18), thereby mediating cellular interactions to lymphocytes. CD102 plays a role in NK cytotoxicity and migration, and lymphocyte recirculation. Expression of CD102 is found on most lymphocytes, monocytes and platelets. The 3C4 (mIC2/4) antibody blocks interaction of lymphocytes with antigen presenting cells. Applications Reported: This 3C4 (mIC2/4) antibody has been reported for use in flow cytometric analysis, and immunohistology staining of frozen tissue sections. Applications Tested: This 3C4 (mIC2/4) antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
CD102 (ICAM-2) Monoclonal antibody specifically detects CD102 (ICAM-2) in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry (Frozen).Specifications
CD102 (ICAM-2) | |
0.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
P35330 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
3C4 (mIC2/4) | |
Flow Cytometry, Immunohistochemistry (Frozen) | |
Alexa Fluor 488 | |
Rat | |
Mouse | |
15896 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! |
For Research Use Only.