missing translation for 'onlineSavingsMsg'
Learn More

CD100 (SEMA4D), Alexa Fluor 488, clone: eBio133-1C6 (133-1C6), eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 53-1009-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15500197

  • £94.39 / 25 tests


Estimated Shipment date
from Supplier 04-04-2018
Add to basket

Description

Description

Description: The eBio133-1C6 monoclonal antibody reacts with human CD100, a member of the class IV semaphorin family also called sema4D. CD100 promotes angiogenesis, invasive growth, proliferation, and anti-apoptosis of cancer cells in vitro. Furthermore CD100 is a costimulatory molecule in adaptive immune responses by switching off negative signalling from CD72. In addition to CD72 another ligand is Plexin-B1. It has been shown that higher expression levels of CD100 are correlated with poor survival in soft tissue sarcoma patients. Expression is found in both hematopoietic and non-hematopoietic cells.CD100 is expressed on resting T cells and NK cells and B cell. CD100 can also exist as a soluble form.Applications Reported: This eBio133-1C6 (133-1C6) antibody has been reported for use in flow cytometric analysis.Applications Tested: This eBio133-1C6 (133-1C6) antibody has been pre-titrated and tested by flow cytometric of human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD100 (SEMA4D)
eBio133-1C6 (133-1C6)
Alexa Fluor 488
Conjugated
Murine
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
5μL (0.25μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Semaphorin 4D
IgM
RUO
Primary
Human
Videos

For Research Use Only.