missing translation for 'onlineSavingsMsg'
Learn More

CD10, FITC, clone: eBioCB-CALLA (CB-CALLA), eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-0106-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15516336

  • £56.98 / 25 tests


Estimated Shipment date
from Supplier 20-02-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The eBioCB-CALLA monoclonal antibody recognizes human CD10 (CALLA, NEP, enkephalinase, Neprilysin), which is a 100 kDa, type II cell surface glycoprotein originally identified for its expression on most acute lymphoblastic leukemias (ALL). Subsequently, CD10 was shown to be the same molecule as the neutral endopeptidase (NEP), or KII-NA. CD10 is a Zn2+-dependent metallo-peptidase with endothelin, glucagon, gastrin, neurotensin and bradykinin included among its substrates. CD10 is involved in the regulation of chemotactic and inflammatory processes involving neutrophils. In B cells, CD10 regulates stromal cell-dependent B lymphopoiesis and expression has also been reported on mature B cells in germinal centres. In addition to the hematopoietic compartment, other major sites of CD10 expression are the brush border of enterocytes and renal tubules and glomeruli. There is partial blocking of the eBioCB-CALLA and MEM-78 monoclonal antibodies indicating that they recognize similar epitopes.Applications Reported: This eBioCB-CALLA (CB-CALLA) antibody has been reported for use in flow cytometric analysis.Applications Tested: This eBioCB-CALLA (CB-CALLA) antibody has been pre-titrated and tested by flow cytometric analysis of normal normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD10
eBioCB-CALLA (CB-CALLA)
FITC
Conjugated
Murine
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
5μL (0.125μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Common Acute Lymphocytic Leukemia antigen (CALLA), Neprilysin
IgG2b
RUO
Primary
Human
Videos