missing translation for 'onlineSavingsMsg'
Learn More

CD 2665, Tocris Bioscience™

Selective RARβ/γ antagonist

Brand:  Tocris (Bio-Techne) 380010

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13433624

  • £219.03 / 10mg

Estimated Shipment date
from Supplier 26-02-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective RARβγ antagonist (KD values are 110, 306 and > 1000 nM for RARγ, RARβ and RARα respectively). Blocks retinoic acid-induced apoptosis ex vivo.
Specifications

Chemical Identifiers

170355-78-9
486.608
4-7-1-adamantyl-6-2-methoxyethoxymethoxy naphthalen-2-yl benzoic acid, 4-6-2-methoxyethoxy methoxy-7-tricyclo 3.3.1.13,7 dec-1-yl-2-naphthalenyl benzoic acid, d0y6wj, 4-7-adamantan-1-yl-6-2-methoxyethoxy methoxy-2-naphthyl benzoic acid, 4-6-2-methoxyethoxy methoxy-7-tricyclo 3.3.1.1.3,7 dec-1-yl-2-naphthalenyl benzoic acid, 4-7-1-adamantyl-6-methoxyethoxymethoxy-2-naphthyl benzoic acid, benzoic acid, 4-6-2-methoxyethoxy methoxy-7-tricyclo 3.3.1.1.3,7 dec-1-yl-2-naphthalenyl, benzoic acid,4-6-2-methoxyethoxy methoxy-7-tricyclo 3.3.1.13,7 dec-1-yl-2-naphthalenyl
4-[7-(1-adamantyl)-6-(2-methoxyethoxymethoxy)naphthalen-2-yl]benzoic acid
C31H34O5
JBALRFFXKQPVLT-UHFFFAOYSA-N
216241
COCCOCOC1=C(C=C2C=C(C=CC2=C1)C3=CC=C(C=C3)C(=O)O)C45CC6CC(C4)CC(C6)C5

Specifications

10mg
170355-78-9
4-7-1-adamantyl-6-2-methoxyethoxymethoxy naphthalen-2-yl benzoic acid, 4-6-2-methoxyethoxy methoxy-7-tricyclo 3.3.1.13,7 dec-1-yl-2-naphthalenyl benzoic acid, d0y6wj, 4-7-adamantan-1-yl-6-2-methoxyethoxy methoxy-2-naphthyl benzoic acid, 4-6-2-methoxyethoxy methoxy-7-tricyclo 3.3.1.1.3,7 dec-1-yl-2-naphthalenyl benzoic acid, 4-7-1-adamantyl-6-methoxyethoxymethoxy-2-naphthyl benzoic acid, benzoic acid, 4-6-2-methoxyethoxy methoxy-7-tricyclo 3.3.1.1.3,7 dec-1-yl-2-naphthalenyl, benzoic acid,4-6-2-methoxyethoxy methoxy-7-tricyclo 3.3.1.13,7 dec-1-yl-2-naphthalenyl
COCCOCOC1=C(C=C2C=C(C=CC2=C1)C3=CC=C(C=C3)C(=O)O)C45CC6CC(C4)CC(C6)C5
486.608
486.6
CD 2665
C31H34O5
JBALRFFXKQPVLT-UHFFFAOYSA-N
4-[7-(1-adamantyl)-6-(2-methoxyethoxymethoxy)naphthalen-2-yl]benzoic acid
216241
>98%
Videos
Documents