missing translation for 'onlineSavingsMsg'
Learn More

CD 1530, Tocris Bioscience™

Potent and selective RARγ agonist

Brand:  Tocris (Bio-Techne) 255450

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13461454

  • £569.42 / 50mg


Estimated Shipment
07-03-2019
Add to basket

Description

Description

Potent and selective RARγ receptor agonist (Ki values are 150, 1500 and 2750 nM for RARγ, RARβ and RARα receptors respectively). Activates transcriptional activity (AC50 = 1.8 nM).
Specifications

Chemical Identifiers

107430-66-0
398.502
cd hplc
4-[7-(1-adamantyl)-6-hydroxynaphthalen-2-yl]benzoic acid
C27H26O3
VCQGNUWOMLYNNG-UHFFFAOYSA-N
9952709
C1C2CC3CC1CC(C2)(C3)C4=C(C=C5C=CC(=CC5=C4)C6=CC=C(C=C6)C(=O)O)O

Specifications

CD 1530
107430-66-0
cd hplc
C1C2CC3CC1CC(C2)(C3)C4=C(C=C5C=CC(=CC5=C4)C6=CC=C(C=C6)C(=O)O)O
398.502
398.49
50mg
C27H26O3
VCQGNUWOMLYNNG-UHFFFAOYSA-N
4-[7-(1-adamantyl)-6-hydroxynaphthalen-2-yl]benzoic acid
9952709
>98%
Videos