Learn More
CCMQ, Tocris Bioscience™
Used to characterize NR2B-containing NMDA receptors
Brand:  Tocris (Bio-Techne) 123850
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13428943
  • £389.55 / 50mg
Estimated Shipment date
from Supplier 01-04-2020
Description
Description
For use in conjunction with [3H]-homoquinolinic acid to characterize GluN2B (formally NR2B) containing NMDA receptors; selectively inhibits [3H]-homoquinolinic acid binding to non-NMDA sensitive sites.
Please refer to IUPHAR Guide to Pharmacology for the most recent naming conventions.
Specifications
Chemical Identifiers
132623-44-0
231.207
ccmq, 2-carboxy-3-carboxymethylquinoline, chembl25599, 3-quinolineacetic acid, 2-carboxy, tocris-1238, 2-carboxy-3-quinolineacetic acid, 3-carboxymethyl-quinoline-2-carboxylic acid, 3-carboxymethyl quinoline-2-carboxylic acid
3-(carboxymethyl)quinoline-2-carboxylic acid
C12H9NO4
IFXJDEHFADWURH-UHFFFAOYSA-N
3337816
C1=CC=C2C(=C1)C=C(C(=N2)C(=O)O)CC(=O)O
Specifications
50mg
132623-44-0
ccmq, 2-carboxy-3-carboxymethylquinoline, chembl25599, 3-quinolineacetic acid, 2-carboxy, tocris-1238, 2-carboxy-3-quinolineacetic acid, 3-carboxymethyl-quinoline-2-carboxylic acid, 3-carboxymethyl quinoline-2-carboxylic acid
C1=CC=C2C(=C1)C=C(C(=N2)C(=O)O)CC(=O)O
231.207
231.21
CCMQ
C12H9NO4
IFXJDEHFADWURH-UHFFFAOYSA-N
3-(carboxymethyl)quinoline-2-carboxylic acid
3337816
>98%