missing translation for 'onlineSavingsMsg'
Learn More

CCL8 (MCP-2) Mouse anti-Human, eFluor® 660, Clone: DWZEE, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 50-9789-41

Code : NEW

Additional Details : Weight : 0.23750kg

Product Code. 15580137

  • £72.50 / 25 tests

Estimated Shipment
04-11-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This can be used at 5 µL (0.125 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. eFluor® 660 is a replacement for Alexa Fluor® 647. eFluor® 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered. This gene is one of several cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The protein encoded by this gene is structurally related to the CXC subfamily of cytokines. Members of this subfamily are characterized by two cysteines separated by a single amino acid. This cytokine displays chemotactic activity for monocytes, lymphocytes, basophils and eosinophils. By recruiting leukocytes to sites of inflammation this cytokine may contribute to tumor-associated leukocyte infiltration and to the antiviral state against HIV infection.
Specifications

Specifications

CCL8 (MCP-2)
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P80075
HC14, MCP-2, MCP2, SCYA10, SCYA8
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
DWZEE
eFluor™ 660
CCL8
Liquid
CCL8
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
6355
Videos
Documents

For Research Use Only.