missing translation for 'onlineSavingsMsg'
Learn More

CCL2 (MCP-1) Armenian Hamster anti-Human, Mouse, Rat, FITC, Clone: 2H5, eBioscience™

Armenian Hamster Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-7096-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15586586

  • £59.25 / 25 tests

Estimated Shipment
25-11-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene is one of several cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The protein encoded by this gene is structurally related to the CXC subfamily of cytokines. Members of this subfamily are characterized by two cysteines separated by a single amino acid. This cytokine displays chemotactic activity for monocytes and basophils but not for neutrophils or eosinophils. It has been implicated in the pathogenesis of diseases characterized by monocytic infiltrates, like psoriasis, rheumatoid arthritis and atherosclerosis. It binds to chemokine receptors CCR2 and CCR4.
Specifications

Specifications

CCL2 (MCP-1)
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P13500, P10148, P14844
GDCF-2, HC11, HSMCR30, MCAF, MCP-1, MCP1, MGC9434, SCYA2, SMC-CF
Armenian Hamster
Affinity chromatography
RUO
Antibody
Monoclonal
Human, Mouse, Rat
Flow Cytometry
2H5
FITC
CCL2
Liquid
CCL2
IgG
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
20296, 24770, 6347
Videos
Documents

For Research Use Only.