missing translation for 'onlineSavingsMsg'
Learn More

Human CCL11/Eotaxin Antibody, R&D Systems™

Mouse Monoclonal Antibody

POA

Specifications

Antigen CCL11/Eotaxin
Clone 43915
Dilution Western Blot 1 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
View More Specs

Description

Description

CCL11/Eotaxin Monoclonal specifically detects CCL11/Eotaxin in Human samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready.
Specifications

Specifications

CCL11/Eotaxin
Western Blot 1 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
6356
E. coli-derived recombinant human CCL11/Eotaxin
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human CCL11/Eotaxin in Western blots. Does not cross-react with recombinant cotton rat CCL3, 4, 5, recombinant human CCL1, 3, 4, 5, 8, 14, 15, 16, 17, 18, 19, 20, 21, 22, 23, 24, 25, 26, 27, recombinant mouse CCL1, 2, 3, 4, 5, 6, 7, 8, 9/10/MIP-1 gamma , 11, 12, 17, 19, 20, 21, 22, 24, 25, 27, or recombinant rat CCL20.
43915
Western Blot, Flow Cytometry, CyTOF
Unconjugated
Mouse
Human
C-C motif chemokine 11, chemokine (C-C motif) ligand 11, Eosinophil chemotactic protein, Eotaxin, eotaxin-1, MGC22554, SCYA11Small-inducible cytokine A11, small inducible cytokine subfamily A (Cys-Cys), member 11 (eotaxin)
CCL11
IgG1
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only