missing translation for 'onlineSavingsMsg'
Learn More

Motic CCIS™ Plan Achromatic Objective UC

Color corrected infinity optical system/plan achromat objective for crisp, flat, high-contrast images.

Brand:  Motic 1101001705221

Additional Details : Weight : 0.20000kg

 View more versions of this product

Product Code. 15940571

  • £88.81 / Each


Estimated Shipment
17-05-2021
Add to basket

Description

Description

  • Color corrected infinity system.
    • Brilliant, flawless image quality.
    • Image aberrations are corrected right to the edge of the field of view.
  • Plan achromat lenses.
    • Provide field flatness.
  • Suitable for the following:
    • Panthera U Binocular
    • Panthera U Trinocular
    • Panthera KU Binocular
    • Panthera KU Trinocular
    • Panthera C Binocular
    • Panthera C Trinocular
    • Panthera CC Binocular
    • Panthera CC Trinocular
    • Panthera DL
    • Panthera L
Specifications

Specifications

Objective mounting thread: W 4/5 in x 1/36 in. (RMS standard)
Objective NA: 0.45, WD: 0.80 mm, DIN: 45 mm
Objective tube length: Infinity CCIS™
Panthera series microscopes
Objective type: Plan achromatic
20X
Videos
SDS
Documents