missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Viral CCI Antibody, R&D Systems™
Mouse Monoclonal Antibody
POA
Specifications
Antigen | CCI |
---|---|
Clone | 112811 |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
CCI Monoclonal specifically detects CCI in Viral samples. It is validated for Western Blot.Specifications
CCI | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant viral CCI | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects viral CCI in direct ELISAs and Western blots. |
112811 | |
Western Blot | |
Unconjugated | |
Mouse | |
Virus | |
3707616 | |
IgG1 | |
Protein A or G purified from ascites | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only