missing translation for 'onlineSavingsMsg'
Learn More

CCG 50014, Tocris Bioscience™

Potent and selective inhibitor of regulator of G-protein signaling 4 (RGS4) protein

Brand:  RD Systems (Bio-Techne) 4567/50

Code : Z1

Additional Details : Weight : 0.00050kg

Quantity 50mg

 View more versions of this product

Product Code. 13464934

  • £563.01 / 50mg

Estimated Shipment date
from Supplier 31-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent and selective inhibitor of regulator of G-protein signaling 4 (RGS4) protein (IC50 values are 30.1 nM and 11.0 μM for RGS4 and RGS8 respectively). Exhibits >20-fold selectivity for RGS4 over other RGS proteins.
Specifications

Chemical Identifiers

883050-24-6
316.35
compound 1a, unii-ra72g28ve9, 4-4-fluorobenzyl-2-p-tolyl-1,2,4-thiadiazolidine-3,5-dione, 4-4-fluorophenyl methyl-2-4-methylphenyl-1,2,4-thiadiazolidine-3,5-dione, cyto9f11, maybridge1_007327, d0c2gr, 4-4-fluorobenzyl-2-4-methylphenyl-1,2,4-thiadiazolane-3,5-dione, 4-4-fluorophenyl methyl-2-p-tolyl-1,2,4-thiadiazolidine-3,5-dione
4-[(4-fluorophenyl)methyl]-2-(4-methylphenyl)-1,2,4-thiadiazolidine-3,5-dione
C16H13FN2O2S
QUIIIYITNGOFEI-UHFFFAOYSA-N
2733079
CC1=CC=C(C=C1)N2C(=O)N(C(=O)S2)CC3=CC=C(C=C3)F

Specifications

50mg
883050-24-6
compound 1a, unii-ra72g28ve9, 4-4-fluorobenzyl-2-p-tolyl-1,2,4-thiadiazolidine-3,5-dione, 4-4-fluorophenyl methyl-2-4-methylphenyl-1,2,4-thiadiazolidine-3,5-dione, cyto9f11, maybridge1_007327, d0c2gr, 4-4-fluorobenzyl-2-4-methylphenyl-1,2,4-thiadiazolane-3,5-dione, 4-4-fluorophenyl methyl-2-p-tolyl-1,2,4-thiadiazolidine-3,5-dione
CC1=CC=C(C=C1)N2C(=O)N(C(=O)S2)CC3=CC=C(C=C3)F
316.35
316.35
CCG 50014
C16H13FN2O2S
QUIIIYITNGOFEI-UHFFFAOYSA-N
4-[(4-fluorophenyl)methyl]-2-(4-methylphenyl)-1,2,4-thiadiazolidine-3,5-dione
2733079
>98%
Videos
Documents