missing translation for 'onlineSavingsMsg'
Learn More

CB 13, Tocris Bioscience™

Potent dual CB1/CB2 agonist

Brand:  Tocris (Bio-Techne) 292810

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13442064

  • £144.06 / 10mg

Estimated Shipment date
from Supplier 20-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent, orally active CB1/CB2 receptor agonist with limited brain penetration (EC50 values are 6.1 and 27.9 nM for CB1 and CB2 receptors respectively). Displays antihyperalgesic activity in a rat model of neuropathic pain with no CNS side effects.
Specifications

Chemical Identifiers

432047-72-8
368.476
1-naphthalenyl 4-pentyloxy-1-naphthalenyl methanone, cra-13, unii-9xrj6055xt, naphthalen-1-yl-4-pentyloxynaphthalen-1-yl methanone, naphthalen-1-yl 4-pentyloxy naphthalen-1-yl methanone, cra13, 1-naphthyl-4-pentoxy-1-naphthyl methanone, naphthalen-1-yl-4-pentyloxy-naphthalen-1-yl-methanone, methanone, 1-naphthalenyl 4-pentyloxy-1-naphthalenyl
naphthalen-1-yl-(4-pentoxynaphthalen-1-yl)methanone
C26H24O2
RSUMDJRTAFBISX-UHFFFAOYSA-N
9799417
CCCCCOC1=CC=C(C2=CC=CC=C21)C(=O)C3=CC=CC4=CC=CC=C43

Specifications

10mg
432047-72-8
C26H24O2
RSUMDJRTAFBISX-UHFFFAOYSA-N
naphthalen-1-yl-(4-pentoxynaphthalen-1-yl)methanone
9799417
CB 13
>99%
1-naphthalenyl 4-pentyloxy-1-naphthalenyl methanone, cra-13, unii-9xrj6055xt, naphthalen-1-yl-4-pentyloxynaphthalen-1-yl methanone, naphthalen-1-yl 4-pentyloxy naphthalen-1-yl methanone, cra13, 1-naphthyl-4-pentoxy-1-naphthyl methanone, naphthalen-1-yl-4-pentyloxy-naphthalen-1-yl-methanone, methanone, 1-naphthalenyl 4-pentyloxy-1-naphthalenyl
CCCCCOC1=CC=C(C2=CC=CC=C21)C(=O)C3=CC=CC4=CC=CC=C43
368.476
368.47
Videos
Documents