missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CATSPER4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA550373
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total CATSPER4 protein.
CATSPER4 Polyclonal antibody specifically detects CATSPER4 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
CATSPER4 | |
Polyclonal | |
Unconjugated | |
CATSPER4 | |
4933412L05; cation channel of sperm 4; cation channel sperm associated 4; cation channel sperm-associated protein 4; cation channel, sperm associated 4; CatSper4; channel, sperm associated 4; CTSR4; ion channel CatSper4; RGD1565213 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
378807 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
2.2 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q7RTX7 | |
CATSPER4 | |
Fusion protein corresponding to residues near the C terminal of human cation channel, sperm associated 4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |