missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CATSPER1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA550372
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total CATSPER1 protein.
CATSPER1 Polyclonal antibody specifically detects CATSPER1 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
CATSPER1 | |
Polyclonal | |
Unconjugated | |
CATSPER1 | |
cation channel of sperm 1; cation channel sperm associated 1; cation channel sperm-associated protein 1; cation channel, sperm associated 1; CATSPER; CATSPER1; hCatSper; KSper; sperm ion channel; sperm protein CATSPER1; sperm-associated cation channel 1; SPGF7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
117144 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.8 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q8NEC5 | |
CATSPER1 | |
Fusion protein corresponding to residues near the C terminal of human cation channel, sperm associated 1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |