missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cathepsin S Antibody, R&D Systems™
Rat Monoclonal Antibody
POA
Specifications
Antigen | Cathepsin S |
---|---|
Clone | 248718 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunoprecipitation |
Description
Cathepsin S Monoclonal specifically detects Cathepsin S in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Cathepsin S | |
LYOPH | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rat | |
Human | |
P25774 | |
1520 | |
Mouse myeloma cell line NS0-derived recombinant human Cathepsin S Gln17-Ile331 (predicted) Accession # P25774 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects pro and mature forms of human Cathepsin S in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human Cathepsin B, C, D, L, L2/V, X/Z/P, recombinant mouse Cathepsin F, or H is observed. |
248718 | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
cathepsin S, CTSS, EC 3.4.22, EC 3.4.22.27, FLJ50259, MGC3886 | |
CTSS | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only