missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cathepsin O Antibody, R&D Systems™
Goat Polyclonal Antibody
POA
Specifications
Antigen | Cathepsin O |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunohistochemistry, Immunoprecipitation |
Classification | Polyclonal |
Description
Cathepsin O Polyclonal specifically detects Cathepsin O in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.Specifications
Cathepsin O | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL | |
Polyclonal | |
Goat | |
Human | |
cathepsin O, CTSO, CTSO1, EC 3.4.22.42 | |
CTSO | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunohistochemistry, Immunoprecipitation | |
Unconjugated | |
RUO | |
P43234 | |
1519 | |
Mouse myeloma cell line NS0-derived recombinant human Cathepsin O Asp24-Val321 Accession # P43234 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human Cathepsin O in direct ELISAs and Western blots. In Western blots, less than 2% cross-reactivity with recombinant human Cathepsin A, B, C, D, L, and Z is observed. |
For Research Use Only