missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cathepsin L Mouse anti-Insect, Clone: 193702, R&D Systems™
Mouse Monoclonal Antibody has been used in 7 publications
POA
Specifications
Antigen | Cathepsin L |
---|---|
Clone | 193702 |
Immunogen | S. frugiperda ovarian cell line Sf 21-derived Insect Cathepsin L |
Gene ID (Entrez) | 1514 |
Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
Description
Cathepsin L Monoclonal specifically detects Cathepsin L in Insect samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Cathepsin L | |
S. frugiperda ovarian cell line Sf 21-derived Insect Cathepsin L | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Primary | |
Detects pro and mature forms of Insect Cathepsin L in direct ELISAs and Western blots. In Western blots, less than 5% cross-reactivity with recombinant human Cathepsin L is observed and no cross-reactivity with recombinant mouse Cathepsin L is observed. | |
Unconjugated | |
Western Blot, Immunoprecipitation | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
193702 | |
1514 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Mouse | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
RUO |
For Research Use Only