missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Cathepsin E Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 1 publication
POA
Specifications
Antigen | Cathepsin E |
---|---|
Clone | 186513 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
Cathepsin E Monoclonal specifically detects Cathepsin E in Mouse samples. It is validated for Western Blot.Specifications
Cathepsin E | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
P70269 | |
1510 | |
Mouse myeloma cell line NS0-derived recombinant mouse Cathepsin E Gln19-Pro397 Accession # P70269 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse Cathepsin E in direct ELISAs and Western blots. It recognizes both the pro and active forms of recombinant mouse (rm) Cathepsin E. In Western blots, no cross-reactivity with rmCathepsin B, C, H, L, or recombinant human Cathepsin O, S, or V is observed. |
186513 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CATE, cathepsin E, CTSE, EC 3.4.23, EC 3.4.23.34, erythrocyte membrane aspartic proteinase, slow-moving proteinase | |
CTSE | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only