missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Cathepsin A/Lysosomal Carboxypeptidase A Antibody, R&D Systems™
Rat Monoclonal Antibody
POA
Specifications
Antigen | Cathepsin A/Lysosomal Carboxypeptidase A |
---|---|
Clone | 191424 |
Concentration | LYOPH |
Dilution | Immunohistochemistry 8-25 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Immunohistochemistry, Immunoprecipitation |
Description
Cathepsin A/Lysosomal Carboxypeptidase A Monoclonal specifically detects Cathepsin A/Lysosomal Carboxypeptidase A in Mouse samples. It is validated for Immunohistochemistry, Immunoprecipitation.Specifications
Cathepsin A/Lysosomal Carboxypeptidase A | |
LYOPH | |
Immunohistochemistry, Immunoprecipitation | |
Unconjugated | |
Rat | |
Mouse | |
beta-galactosidase 2, beta-galactosidase protective protein, Carboxypeptidase C, Carboxypeptidase L, cathepsin ANGBE, CTSA, EC 3.4.16, EC 3.4.16.5, GLB2, GSL, Lysosomal Carboxypeptidase A, lysosomal protective protein, PPCA, PPGBprotective protein for beta-galactosidase (galactosialidosis), Protective protein cathepsin A, Protective protein for beta-galactosidase | |
CTSA | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
191424 | |
Immunohistochemistry 8-25 ug/mL, Immunoprecipitation 25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
5476 | |
Mouse myeloma cell line NS0-derived recombinant mouse Cathepsin A aa 24-474 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse Cathepsin A in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Cathepsin A or recombinant mouse DPPIV/CD26 is observed. |
For Research Use Only