missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Cathepsin 7/Cathepsin 1 Antibody, R&D Systems™
Rat Monoclonal Antibody
POA
Specifications
Antigen | Cathepsin 7/Cathepsin 1 |
---|---|
Clone | 228905 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunoprecipitation |
Description
Cathepsin 7/Cathepsin 1 Monoclonal specifically detects Cathepsin 7/Cathepsin 1 in Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Cathepsin 7/Cathepsin 1 | |
LYOPH | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rat | |
Mouse | |
Q91ZF2 | |
Mouse myeloma cell line NS0-derived recombinant mouse Cathepsin 7/Cathepsin 1 Ala18-Leu331 Accession # Q91ZF2 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse Cathepsin 7/Cathepsin 1 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human Cathepsin O, S, or V, or recombinant mouse Cathepsin B, C, H, L, or Z is observed. |
228905 | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Cts7 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only