missing translation for 'onlineSavingsMsg'
Learn More

Cathepsin 3 Rat anti-Mouse, Clone: 264013, R&D Systems™

Rat Monoclonal Antibody

£136.16 - £390.72

Specifications

Antigen Cathepsin 3
Clone 264013
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Cathepsin 3 Ser18-Val332 Accession # NP_081182
Gene ID (Entrez) 117066
Gene Accession No. NP_081182
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15755474 View Documents RD Systems (Bio-Techne)
MAB2638-SP
25 ug £136.16
25µg View alternative products
Save Up to 
»
Estimated Shipment
13-07-2020
Please to purchase this item. Need a web account? Register with us today!
13459595 View Documents RD Systems (Bio-Techne)
MAB2638
500 ug £390.72
500µg View alternative products
Save Up to 
»
Estimated Shipment
10-08-2020
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cathepsin 3 Monoclonal antibody MAB2638SP Specifically detects Cathepsin 3 in Mouse samples. It is validated for Immunoprecipitation, Western Blotting
Specifications

Specifications

Cathepsin 3
Mouse myeloma cell line NS0-derived recombinant mouse Cathepsin 3 Ser18-Val332 Accession # NP_081182
NP_081182
Reconstitute at 0.5 mg/mL in sterile PBS.
Primary
Detects mouse Cathepsin 3 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant mouse Cathepsin 1, 2, or 6 is observed.
Unconjugated
Purified
RUO
264013
117066
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Monoclonal
Rat
IgG2b
Protein A or G purified from hybridoma culture supernatant
Western Blot, Immunoprecipitation
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.

For Research Use Only