missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Lab Vision™ β-Catenin Rabbit Monoclonal Antibody
Ensure accurate and reproducible results in immunohistochemistry applications with this antibody, part of a comprehensive antibody portfolio.
£162.18 - £800.00
Specifications
Antigen | Catenin, beta |
---|---|
Clone | E247 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
12683597
|
Epredia™
RM-2101-S |
Concentrated | 1 mL |
£800.00
1mL |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12693597
|
Epredia™
RM-2101-RQ |
Ready-to-Use | 12 mL |
£560.00
12mL |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12673597
|
Epredia™
RM-2101-R7 |
Prediluted | 7 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12603607
|
Epredia™
RM-2101-S0 |
Concentrated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
The catenins (α, β and γ) are ubiquitously expressed cytoplasmic proteins associated with E-cadherin at cellular junctions. β-catenin also binds to N-cadherin and co-immunoprecipitates with APC. Cadherin/catenin complexes are linked to the cytoskeleton via a direct association between α-actinin and α-catenin. Increased tyrosine phosphorylation can disrupt catenin-cadherin complexes, influencing cellular adhesion. Mutations in the catenin gene, CTNNB1, have been implicated in its accumulation and various forms of carcinomas.Host Species: Rabbit
Clone: E247
Species Reactivity: Human. Others not known.
Immunogen: A synthetic peptide derived from near the N-terminus of human β-catenin
Molecular Weight: 92 kDa
Positive Control: Breast CA
Cellular Localization: Cell Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Catenin, beta | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
IVD | |
A synthetic peptide derived from near the N-terminus of human beta-catenin |
E247 | |
Monoclonal | |
Rabbit | |
Cancer and Tumor Biology | |
Primary |