Learn More
Catechol, Extra Pure, SLR, Fisher Chemical™
Brand: Fisher Chemical C/3320/48
6.80 GBP valid until 2022-05-29
Use promo code "17501" to get your promotional price.
Code : 21
Additional Details : CAS Number : 120-80-9 Weight : 0.10000kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III
Quantity | 100g |
---|
Description
Catechol, Extra Pure, SLR
Chemical Identifiers
120-80-9 | |
Brown | |
103°C | |
2188 | |
Plastic powder jar |
245°C | |
110.11g/mol | |
6 | |
C6H6O2 |
Specifications
120-80-9 | |
Brown | |
245°C | |
2188 | |
100g | |
Solid |
6 | |
103°C | |
C6H6O2 | |
Plastic powder jar | |
110.11g/mol |