missing translation for 'onlineSavingsMsg'
Learn More

Cataxa/Rh(COD)/(S,S-Dipamp), Thermo Scientific™

Catalog Number 11306239
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
MFCD08064220
This item is not returnable. View return policy
MFCD08064220

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • This product is a Rhodium catalyst anchored on alumina via phosphotungstic acid
  • Used in enantioselective hydrogenation of prochiral olefinic bonds
TRUSTED_SUSTAINABILITY
Quantity 1 g
UN Number UN3260
Physical Form Powder
Chemical Name or Material Cataxa/Rh (COD)/ (S,S-Dipamp)
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'